missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromobenzyl bromide, 99%

Catalog No. AAA1272614
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1272614 25 g
AAA1272603 1 g
AAA1272606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1272614 Supplier Thermo Scientific Chemicals Supplier No. A1272614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 56908-88-4 | C7H5Br3 | 328.83 g/mol

3,5-Dibromobenzyl bromide, is an important raw material and intermediateintermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dibromobenzyl bromide, is an important raw material and intermediateintermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in toluene

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to water, oxidizing agents, bases, alcohols, amines, steel.

Chemical Identifiers

CAS 56908-88-4
Molecular Formula C7H5Br3
Molecular Weight (g/mol) 328.83
MDL Number MFCD00052415
InChI Key PWTFRUXTAFBWBW-UHFFFAOYSA-N
Synonym 3,5-dibromobenzyl bromide, 1,3-dibromo-5-bromomethyl benzene, benzene, 1,3-dibromo-5-bromomethyl, alpha,3,5-tribromotoluene, acmc-20andg, 3,5-dibromobenzylbromide, 3,5-dibromo benzyl bromide, 1,3-bis bromanyl-5-bromomethyl benzene
PubChem CID 143427
IUPAC Name 1,3-dibromo-5-(bromomethyl)benzene
SMILES BrCC1=CC(Br)=CC(Br)=C1

Specifications

Melting Point 93°C to 95°C
Quantity 25 g
UN Number UN1759
Beilstein 3237827
Solubility Information Soluble in toluene
Formula Weight 328.84
Percent Purity 99%
Chemical Name or Material 3,5-Dibromobenzyl bromide
Hazard Category H228-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only