missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromobenzaldehyde 97.0+%, TCI America™

Catalog No. D28325G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28325G 5 g
D283225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28325G Supplier TCI America Supplier No. D28325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 56990-02-4
Molecular Formula C7H4Br2O
Molecular Weight (g/mol) 263.916
MDL Number MFCD00156887
InChI Key ZLDMZIXUGCGKMB-UHFFFAOYSA-N
Synonym 3,5-dibromo-benzaldehyde, benzaldehyde, 3,5-dibromo, pubchem3069, acmc-209lvd, ksc493o2p, 3,5-bis bromanyl benzaldehyde, paragos 530307, 3,5-dibromo-phenyl-methanone
PubChem CID 622077
IUPAC Name 3,5-dibromobenzaldehyde
SMILES C1=C(C=C(C=C1Br)Br)C=O

Specifications

Melting Point 93°C
Quantity 5 g
Formula Weight 263.92
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibromobenzaldehyde