missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromobenzaldehyde, 98+%

Catalog No. AC393150050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC393150050 5 g Glass bottle
AC393150250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC393150050 Supplier Thermo Scientific Chemicals Supplier No. 393150050

CAS: 56990-02-4 | C7H4Br2O | 263.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56990-02-4
Molecular Formula C7H4Br2O
Molecular Weight (g/mol) 263.91
MDL Number MFCD00156887
InChI Key ZLDMZIXUGCGKMB-UHFFFAOYSA-N
Synonym 3,5-dibromo-benzaldehyde, benzaldehyde, 3,5-dibromo, pubchem3069, acmc-209lvd, ksc493o2p, 3,5-bis bromanyl benzaldehyde, paragos 530307, 3,5-dibromo-phenyl-methanone
PubChem CID 622077
IUPAC Name 3,5-dibromobenzaldehyde
SMILES C1=C(C=C(C=C1Br)Br)C=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 86°C to 90°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula Br2C6H3C(=O)H
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dmso,ethyl acetate,methanol
Formula Weight 263.91
Percent Purity 98+%
Physical Form Crystals or Powder
Chemical Name or Material 3, 5-Dibromobenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
Recommended Storage 4°C

RUO – Research Use Only