missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromoanthranilic acid, 98+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 393160250

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC393160250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

609-85-8
294.93
WNABMWFLKQEGCP-UHFFFAOYSA-N
219491
C1=C(C=C(C(=C1Br)N)C(=O)O)Br
C7H5Br2NO2
MFCD00017087
3,5-dibromoanthranilic acid, 2-amino-3,5-dibromobenzoicacid, 3,5-dibromo-2-aminobenzoic acid, buttpark 89\07-50, 2-amino-3,5-dibromo-benzoic acid, benzoic acid, 2-amino-3,5-dibromo, acmc-20ahts, pubchem3720, 2-carboxy-4,6-dibromoaniline, ghl.pd_mitscher_leg0.1128
2-amino-3,5-dibromobenzoic acid

Specifications

3, 5-Dibromoanthranilic acid
98.0
Authentic
Glass bottle
25g
3,5-dibromoanthranilic acid, 2-amino-3,5-dibromobenzoicacid, 3,5-dibromo-2-aminobenzoic acid, buttpark 89\07-50, 2-amino-3,5-dibromo-benzoic acid, benzoic acid, 2-amino-3,5-dibromo, acmc-20ahts, pubchem3720, 2-carboxy-4,6-dibromoaniline, ghl.pd_mitscher_leg0.1128
C1=C(C=C(C(=C1Br)N)C(=O)O)Br
294.93
294.93
609-85-8
100.0
98% min. (HPLC)
C7H5Br2NO2
MFCD00017087
WNABMWFLKQEGCP-UHFFFAOYSA-N
2-amino-3,5-dibromobenzoic acid
219491
98+%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Promotions

Promotions