CAS RN 609-85-8
IUPAC Name:
2-amino-3,5-dibromobenzoic acid
Synonyms:
2-amino-3,5-dibromobenzoic acid
Molecular Weight (g/mol):
294.93
Molecular Formula:
C7H5Br2NO2
InChi Key:
WNABMWFLKQEGCP-UHFFFAOYSA-N
SMILES:
NC1=C(Br)C=C(Br)C=C1C(O)=O
1
–
1
of
1
results
1
2-Amino-3,5-dibromobenzoic acid, Thermo Scientific™
CAS: 609-85-8 Molecular Formula: C7H5Br2NO2 Molecular Weight (g/mol): 294.93 InChI Key: WNABMWFLKQEGCP-UHFFFAOYSA-N Synonym: 3,5-dibromoanthranilic acid,2-amino-3,5-dibromobenzoicacid,3,5-dibromo-2-aminobenzoic acid,buttpark 89\07-50,2-amino-3,5-dibromo-benzoic acid,benzoic acid, 2-amino-3,5-dibromo,acmc-20ahts,pubchem3720,2-carboxy-4,6-dibromoaniline,ghl.pd_mitscher_leg0.1128 PubChem CID: 219491 IUPAC Name: 2-amino-3,5-dibromobenzoic acid SMILES: C1=C(C=C(C(=C1Br)N)C(=O)O)Br