missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dibromoanisole 98.0+%, TCI America™
$82.02 - $315.03
Chemical Identifiers
| CAS | 74137-36-3 |
|---|---|
| Molecular Formula | C7H6Br2O |
| Molecular Weight (g/mol) | 265.932 |
| MDL Number | MFCD02258848 |
| InChI Key | OQZAQBGJENJMHT-UHFFFAOYSA-N |
| Synonym | 3,5-dibromoanisole, 1,3-dibromo-5-methoxy-benzene, benzene, 1,3-dibromo-5-methoxy, pubchem2009, acmc-1bh5r, ksc493q1d, 1.3-dibromo-5-methoxybenzene |
| PubChem CID | 11021812 |
| IUPAC Name | 1,3-dibromo-5-methoxybenzene |
| SMILES | COC1=CC(=CC(=C1)Br)Br |
Chemical Identifiers
| 74137-36-3 | |
| 265.932 | |
| OQZAQBGJENJMHT-UHFFFAOYSA-N | |
| 11021812 | |
| COC1=CC(=CC(=C1)Br)Br |
| C7H6Br2O | |
| MFCD02258848 | |
| 3,5-dibromoanisole, 1,3-dibromo-5-methoxy-benzene, benzene, 1,3-dibromo-5-methoxy, pubchem2009, acmc-1bh5r, ksc493q1d, 1.3-dibromo-5-methoxybenzene | |
| 1,3-dibromo-5-methoxybenzene |
Specifications
| 74137-36-3 | |
| 261°C | |
| MFCD02258848 | |
| 3,5-dibromoanisole, 1,3-dibromo-5-methoxy-benzene, benzene, 1,3-dibromo-5-methoxy, pubchem2009, acmc-1bh5r, ksc493q1d, 1.3-dibromo-5-methoxybenzene | |
| COC1=CC(=CC(=C1)Br)Br | |
| 265.932 | |
| 265.93 | |
| Crystalline Powder |
| 39°C | |
| C7H6Br2O | |
| 5 g | |
| OQZAQBGJENJMHT-UHFFFAOYSA-N | |
| 1,3-dibromo-5-methoxybenzene | |
| 11021812 | |
| ≥98.0% (GC) | |
| 3,5-Dibromoanisole |
Safety and Handling
TSCA : Yes