missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromoanisole 98.0+%, TCI America™

Catalog No. D33645G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33645G 5 g
D336425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33645G Supplier TCI America Supplier No. D33645G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 74137-36-3
Molecular Formula C7H6Br2O
Molecular Weight (g/mol) 265.932
MDL Number MFCD02258848
InChI Key OQZAQBGJENJMHT-UHFFFAOYSA-N
Synonym 3,5-dibromoanisole, 1,3-dibromo-5-methoxy-benzene, benzene, 1,3-dibromo-5-methoxy, pubchem2009, acmc-1bh5r, ksc493q1d, 1.3-dibromo-5-methoxybenzene
PubChem CID 11021812
IUPAC Name 1,3-dibromo-5-methoxybenzene
SMILES COC1=CC(=CC(=C1)Br)Br

Specifications

Melting Point 39°C
Boiling Point 261°C
Quantity 5 g
Formula Weight 265.93
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibromoanisole