missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromo-4-pyridinecarboxaldehyde 97.0+%, TCI America™

Catalog No. d46741g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46741G 1 g
D46745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46741G Supplier TCI America Supplier No. D46741G

Chemical Identifiers

CAS 70201-42-2
Molecular Formula C6H3Br2NO
Molecular Weight (g/mol) 264.904
MDL Number MFCD06410681
InChI Key WPBYVMDYYFWYAY-UHFFFAOYSA-N
Synonym 3,5-Dibromoisonicotinaldehyde, 3,5-Dibromo-4-formylpyridine
PubChem CID 2762998
IUPAC Name 3,5-dibromopyridine-4-carbaldehyde
SMILES C1=C(C(=C(C=N1)Br)C=O)Br

Specifications

Melting Point 120°C
Color Yellow
Quantity 1 g
Formula Weight 264.90
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibromo-4-pyridinecarboxaldehyde