missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromo-4-methylpyridine 98.0+%, TCI America™

Catalog No. D41185G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41185G 5 g
D41181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41185G Supplier TCI America Supplier No. D41185G

Chemical Identifiers

CAS 3430-23-7
Molecular Formula C6H5Br2N
Molecular Weight (g/mol) 250.921
MDL Number MFCD00128859
InChI Key BNCGUATWCKZLLN-UHFFFAOYSA-N
Synonym 3,5-dibromo-4-picoline, 3,5-dibromo-4-methyl-pyridine, pyridine, 3,5-dibromo-4-methyl, 4-methyl-3,5-dibromopyridine, dibromo-3,5-4-methylpyridine, pubchem17566, acmc-20a0hi, ksc495g9j, 3,5-dibromo-4-methyl pyridine
PubChem CID 819338
IUPAC Name 3,5-dibromo-4-methylpyridine
SMILES CC1=C(C=NC=C1Br)Br

Specifications

Melting Point 108°C
Color White
Quantity 5 g
Formula Weight 250.92
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibromo-4-methylpyridine