accessibility menu, dialog, popup

UserName

CAS RN 3430-23-7

IUPAC Name: 3,5-dibromo-4-methylpyridine
Synonyms: 3,5-dibromo-4-methylpyridine
Molecular Weight (g/mol): 250.92
Molecular Formula: C6H5Br2N
InChi Key: BNCGUATWCKZLLN-UHFFFAOYSA-N
SMILES: CC1=C(Br)C=NC=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results