missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromo-4-methylphenol, 97%

Catalog No. AAL0066003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0066003 1 g
AAL0066006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0066003 Supplier Thermo Scientific Chemicals Supplier No. L0066003

CAS: 13979-81-2 | C7H6Br2O | 265.932 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13979-81-2
Molecular Formula C7H6Br2O
Molecular Weight (g/mol) 265.932
MDL Number MFCD00013968
InChI Key AXCQKKVGMZCWPC-UHFFFAOYSA-N
PubChem CID 55457
IUPAC Name 3,5-dibromo-4-methylphenol
SMILES CC1=C(C=C(C=C1Br)O)Br

Specifications

Melting Point 106°C to 110°C
Quantity 1 g
Beilstein 2249692
Synonym 3,5-dibromo-p-cresol, phenol, 3,5-dibromo-4-methyl, 3,5-dibromo-4-methyl-phenol, phenol, dibromomethyl, acmc-20abwf, 3,5-dibromo-p-methylphenol, 3,5-dibromo-4-methyl phenol, phenol,3,5-dibromo-4-methyl, 3,5-bis bromanyl-4-methyl-phenol
Formula Weight 265.94
Percent Purity 97%
Chemical Name or Material 3,5-Dibromo-4-methylphenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 237-763-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only