missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromo-4-methylaniline 97.0+%, TCI America™

Catalog No. D47301G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47301G 1 g
D47305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47301G Supplier TCI America Supplier No. D47301G

Chemical Identifiers

CAS 13194-73-5
Molecular Formula C7H7Br2N
Molecular Weight (g/mol) 264.948
MDL Number MFCD00151806
InChI Key AQZDIKCNODUMNY-UHFFFAOYSA-N
Synonym 3,5-dibromo-p-toluidine, 4-amino-2,6-dibromotoluene, benzenamine, 3,5-dibromo-4-methyl, acmc-1bt4r, ksc495c9l, 3,5-dibrom-4-methyl-anilin, 3,5-dibromo-4-methylphenylamine, 3,5-dibromo-4-methylphenyl amine, benzenamine,3,5-dibromo-4-methyl, 3,5-bis bromanyl-4-methyl-aniline
PubChem CID 7015779
IUPAC Name 3,5-dibromo-4-methylaniline
SMILES CC1=C(C=C(C=C1Br)N)Br

Specifications

Melting Point 90°C
Quantity 1 g
Formula Weight 264.95
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibromo-4-methylaniline