missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromo-4-methylaniline, 99%

Catalog No. AAL1310103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1310103 1 g
AAL1310106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1310103 Supplier Thermo Scientific Chemicals Supplier No. L1310103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13194-73-5 | C7H7Br2N | 264.948 g/mol

3,5-Dibromo-4-methylaniline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dibromo-4-methylaniline is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 13194-73-5
Molecular Formula C7H7Br2N
Molecular Weight (g/mol) 264.948
MDL Number MFCD00151806
InChI Key AQZDIKCNODUMNY-UHFFFAOYSA-N
PubChem CID 7015779
IUPAC Name 3,5-dibromo-4-methylaniline
SMILES CC1=C(C=C(C=C1Br)N)Br

Specifications

Melting Point 90°C to 92°C
Quantity 1 g
Beilstein 2639285
Synonym 3,5-dibromo-p-toluidine, 4-amino-2,6-dibromotoluene, benzenamine, 3,5-dibromo-4-methyl, acmc-1bt4r, ksc495c9l, 3,5-dibrom-4-methyl-anilin, 3,5-dibromo-4-methylphenylamine, 3,5-dibromo-4-methylphenyl amine, benzenamine,3,5-dibromo-4-methyl, 3,5-bis bromanyl-4-methyl-aniline
Solubility Information Insoluble in water.
Formula Weight 264.96
Percent Purity 99%
Chemical Name or Material 3,5-Dibromo-4-methylaniline
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only