missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromo-4-methoxypyridine, 98%

Catalog No. p-7055566
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6417703 1 g
AAH6417706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6417703 Supplier Thermo Scientific Chemicals Supplier No. H6417703

CAS: 650140-84-4 | C6H5Br2NO2 | 282.919 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 650140-84-4
Molecular Formula C6H5Br2NO2
Molecular Weight (g/mol) 282.919
MDL Number MFCD13185524
InChI Key KAYQEYHOGFRTAL-UHFFFAOYSA-N
Synonym 3,5-dibromo-4-methoxypyridine 1-oxide, 3,5-dibromo-4-methoxypyridine-n-oxide, 3,5-dibromo-4-methoxypyridin-1-ium-1-olate, pubchem6614
PubChem CID 2784723
IUPAC Name 3,5-dibromo-4-methoxy-1-oxidopyridin-1-ium
SMILES COC1=C(C=[N+](C=C1Br)[O-])Br

Specifications

Color White
Quantity 1 g
Formula Weight 266.92
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3,5-Dibromo-4-methoxypyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only