CAS RN 650140-84-4
IUPAC Name:
3,5-dibromo-4-methoxypyridin-1-ium-1-olate
Synonyms:
3,5-dibromo-4-methoxypyridin-1-ium-1-olate
Molecular Weight (g/mol):
282.92
Molecular Formula:
C6H5Br2NO2
InChi Key:
KAYQEYHOGFRTAL-UHFFFAOYSA-N
SMILES:
COC1=C(Br)C=[N+]([O-])C=C1Br
1
–
1
of
1
results
1
Promotions Available
3,5-Dibromo-4-methoxypyridine, 98%
3,5-Dibromo-4-methoxypyridine, 98%
CAS: 650140-84-4 Molecular Formula: C6H5Br2NO2 Molecular Weight (g/mol): 282.919 MDL Number: MFCD13185524 InChI Key: KAYQEYHOGFRTAL-UHFFFAOYSA-N Synonym: 3,5-dibromo-4-methoxypyridine 1-oxide,3,5-dibromo-4-methoxypyridine-n-oxide,3,5-dibromo-4-methoxypyridin-1-ium-1-olate,pubchem6614 PubChem CID: 2784723 IUPAC Name: 3,5-dibromo-4-methoxy-1-oxidopyridin-1-ium SMILES: COC1=C(C=[N+](C=C1Br)[O-])Br