missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromo-4-hydroxybenzoic acid, 98%

Catalog No. AAA1836122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1836122 100 g
AAA1836114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1836122 Supplier Thermo Scientific Chemicals Supplier No. A1836122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3337-62-0 | C7H4Br2O3 | 295.914 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3337-62-0
Molecular Formula C7H4Br2O3
Molecular Weight (g/mol) 295.914
MDL Number MFCD00002548
InChI Key PHWAJJWKNLWZGJ-UHFFFAOYSA-N
Synonym bromoxynylbenzoic acid, dbha, benzoic acid, 3,5-dibromo-4-hydroxy, dibrhbz, unii-95z5a9epei, 3,5-dibromo-4-hydroxybenzoate, 3,5-dibromo-4-hydroxy-benzoic acid, 3',5'-dibromo-4-hydroxybenzoic acid, 95z5a9epei, rarechem al bo 0009
PubChem CID 76857
ChEBI CHEBI:1395
IUPAC Name 3,5-dibromo-4-hydroxybenzoic acid
SMILES C1=C(C=C(C(=C1Br)O)Br)C(=O)O

Specifications

Melting Point 269°C to 273°C
Quantity 100 g
Beilstein 2416147
Formula Weight 295.93
Percent Purity 98%
Chemical Name or Material 3,5-Dibromo-4-hydroxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 222-075-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only