missing translation for 'onlineSavingsMsg'
Learn More

3',5'-Dibromo-4'-hydroxyacetophenone 97.0+%, TCI America™

Catalog No. D28245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28245G 5 g
D282425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28245G Supplier TCI America Supplier No. D28245G
Only null left

Chemical Identifiers

CAS 2887-72-1
Molecular Formula C8H6Br2O2
Molecular Weight (g/mol) 293.94
MDL Number MFCD00075779
InChI Key ZNWPTJSBHHIXLJ-UHFFFAOYSA-N
PubChem CID 76158
IUPAC Name 1-(3,5-dibromo-4-hydroxyphenyl)ethan-1-one
SMILES CC(=O)C1=CC(Br)=C(O)C(Br)=C1

Specifications

Color Red-White
Quantity 5 g
Formula Weight 293.94
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′,5′-Dibromo-4′-hydroxyacetophenone