accessibility menu, dialog, popup

UserName

H3COBrOHBr

CAS RN 2887-72-1

IUPAC Name: 1-(3,5-dibromo-4-hydroxyphenyl)ethan-1-one
Synonyms: 1-(3,5-dibromo-4-hydroxyphenyl)ethanone
Molecular Weight (g/mol): 293.94
Molecular Formula: C8H6Br2O2
InChi Key: ZNWPTJSBHHIXLJ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(Br)=C(O)C(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results