missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dibromo-2-methylthiophene 95.0+%, TCI America™
$196.36 - $671.89
Chemical Identifiers
| CAS | 29421-73-6 |
|---|---|
| Molecular Formula | C5H4Br2S |
| Molecular Weight (g/mol) | 255.96 |
| MDL Number | MFCD07781197 |
| InChI Key | OGAJGUIMFMRGRB-UHFFFAOYSA-N |
| PubChem CID | 122373 |
| IUPAC Name | 3,5-dibromo-2-methylthiophene |
| SMILES | CC1=C(Br)C=C(Br)S1 |
Chemical Identifiers
| 29421-73-6 | |
| 255.96 | |
| OGAJGUIMFMRGRB-UHFFFAOYSA-N | |
| 3,5-dibromo-2-methylthiophene |
| C5H4Br2S | |
| MFCD07781197 | |
| 122373 | |
| CC1=C(Br)C=C(Br)S1 |
Specifications
| 29421-73-6 | |
| Yellow | |
| C5H4Br2S | |
| 5 g | |
| CC1=C(Br)C=C(Br)S1 | |
| 255.96 | |
| 255.96 | |
| Liquid |
| -15°C | |
| 230°C | |
| MFCD07781197 | |
| OGAJGUIMFMRGRB-UHFFFAOYSA-N | |
| 3,5-dibromo-2-methylthiophene | |
| 122373 | |
| ≥95.0% (GC) | |
| 3,5-Dibromo-2-methylthiophene |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator