accessibility menu, dialog, popup

UserName

CAS RN 29421-73-6

IUPAC Name: 3,5-dibromo-2-methylthiophene
Synonyms: 3,5-dibromo-2-methylthiophene
Molecular Weight (g/mol): 255.96
Molecular Formula: C5H4Br2S
InChi Key: OGAJGUIMFMRGRB-UHFFFAOYSA-N
SMILES: CC1=C(Br)C=C(Br)S1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results