missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dibromo-2-methylthiophene 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | -15°C |
| Color | Yellow |
| Boiling Point | 230°C |
| Quantity | 5 g |
| Formula Weight | 255.96 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 3,5-Dibromo-2-methylthiophene |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |