missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromo-1-trimethylsilylbenzene 97.0+%, TCI America™

Catalog No. D27245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27245G 5 g
D272425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27245G Supplier TCI America Supplier No. D27245G

Chemical Identifiers

CAS 17878-23-8
Molecular Formula C9H12Br2Si
Molecular Weight (g/mol) 308.09
MDL Number MFCD03844787
InChI Key SZQQTWLBYAKIOT-UHFFFAOYSA-N
Synonym (3,5-Dibromophenyl)trimethylsilane
PubChem CID 626244
IUPAC Name (3,5-dibromophenyl)trimethylsilane
SMILES C[Si](C)(C)C1=CC(Br)=CC(Br)=C1

Specifications

Melting Point 40°C
Color White-Yellow
Quantity 5 g
Formula Weight 308.09
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3,5-Dibromo-1-trimethylsilylbenzene