missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromo-1,2-phenylenediamine Monohydrochloride 99.0+%, TCI America™

Catalog No. A5080500MG
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5080500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5080500MG Supplier TCI America Supplier No. A5080500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sensitive reagent for the determination of Se by GC-ECD]

Chemical Identifiers

CAS 75568-11-5
Molecular Formula C6H7Br2ClN2
Molecular Weight (g/mol) 302.394
MDL Number MFCD00012967
InChI Key QOZDVKFQMGARCC-UHFFFAOYSA-N
Synonym 3,5-dibromo-1,2-phenylenediamine monohydrochloride, 3,5-dibromobenzene-1,2-diamine hydrochloride, acmc-20andn, 1,2-diamino-3,5-dibromobenzene monohydrochloride, 3,5-bis bromanyl benzene-1,2-diamine hydrochloride, 3,5-dibromo-1,2-phenylenediamine hydrochloride, 1,2-benzenediamine,3,5-dibromo-, hydrochloride 1:1, 3,5-dibromo-ortho-phenylenediamine monohydrochloride, 3,5-dibromobenzene-1,2-diamine-hydrogen chloride 1/1, 3,5-dibromo-1,2-phenylenediaminemonohydrochloride sensitivereagentforthedeterminationofsebygc-ecd
PubChem CID 2724285
IUPAC Name 3,5-dibromobenzene-1,2-diamine;hydrochloride
SMILES C1=C(C=C(C(=C1Br)N)N)Br.Cl

Specifications

Color Red-Yellow
Quantity 500 mg
Formula Weight 302.39
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibromo-1,2-phenylenediamine Monohydrochloride [Sensitive reagent for the determination of Se by
TSCA No
Recommended Storage Refrigerator