missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibenzyloxybenzyl Bromide 98.0+%, TCI America™

Catalog No. p-7119474
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20935G 5 g
B209325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20935G Supplier TCI America Supplier No. B20935G

Chemical Identifiers

CAS 24131-32-6
Molecular Formula C21H19BrO2
Molecular Weight (g/mol) 383.285
MDL Number MFCD02093444
InChI Key WGMYJGAUAQXYFQ-UHFFFAOYSA-N
Synonym 3,5-Bis(benzyloxy)benzyl Bromide, 3,5-Dibenzyloxy-alpha-bromotoluene
PubChem CID 2761019
IUPAC Name 1-(bromomethyl)-3,5-bis(phenylmethoxy)benzene
SMILES C1=CC=C(C=C1)COC2=CC(=CC(=C2)CBr)OCC3=CC=CC=C3

Specifications

Melting Point 87°C
Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 383.29
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibenzyloxybenzyl Bromide
TSCA No
Recommended Storage Refrigerator