missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibenzyloxybenzyl Alcohol 98.0+%, TCI America™

Catalog No. p-7127426
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26515G 5 g
D265125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26515G Supplier TCI America Supplier No. D26515G
Only null left

Chemical Identifiers

CAS 24131-31-5
Molecular Formula C21H20O3
Molecular Weight (g/mol) 320.388
MDL Number MFCD01863236
InChI Key MHHXKZKHZMSINU-UHFFFAOYSA-N
Synonym 3,5-dibenzyloxybenzyl alcohol, 3,5-bis benzyloxy phenyl methanol, 3,5-bis benzyloxy benzyl alcohol, 3,5-bis phenylmethoxy phenyl methanol, 3,5-benzyloxybenzyl alcohol, 3,5-bis phenylmethoxy phenyl methan-1-ol, pubchem13641, 3,5-dibenzyloxyphenyl methanol, mhhxkzkhzmsinu-uhfffaoysa
PubChem CID 561476
IUPAC Name [3,5-bis(phenylmethoxy)phenyl]methanol
SMILES C1=CC=C(C=C1)COC2=CC(=CC(=C2)CO)OCC3=CC=CC=C3

Specifications

Melting Point 82°C
Color White
Quantity 5 g
Formula Weight 320.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibenzyloxybenzyl Alcohol