missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diamino-2,4,6-trimethylbenzenesulfonic Acid 93.0+%, TCI America™

Catalog No. D27095G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27095G 5 g
D270925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27095G Supplier TCI America Supplier No. D27095G

Chemical Identifiers

CAS 32432-55-6
Molecular Formula C9H14N2O3S
Molecular Weight (g/mol) 230.282
MDL Number MFCD00043827
InChI Key PKKGGWLTUCMSSD-UHFFFAOYSA-N
Synonym diaminomesitylenesulfonic acid, benzenesulfonic acid, 3,5-diamino-2,4,6-trimethyl, 2,4-diamino-6-sulfomesitylene, 3,5-diamino-2,4,6-trimethylbenzenesulphonic acid, 2,4,6-trimethyl-3,5-diaminobenzenesulfonic acid, 3,5-diamino-2,4,6-trimethylbenzene sulfonic acid, acmc-209ht1, 2-14-00-00453 beilstein handbook reference, 2,6-diaminomesitylene-4-sulfonic acid, 3,5-diamino-2,4,6-trimethylbenzenesulfonicacid
PubChem CID 94430
IUPAC Name 3,5-diamino-2,4,6-trimethylbenzenesulfonic acid
SMILES CC1=C(C(=C(C(=C1N)C)S(=O)(=O)O)C)N

Specifications

Quantity 5 g
Formula Weight 230.28
Percent Purity ≥93.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Diamino-2,4,6-trimethylbenzenesulfonic Acid
EINECSNumber (3)-2063
RTECSNumber DB5987000
TSCA Yes