missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diamino-1,2,4-triazole 97.0+%, TCI America™

Catalog No. p-7127382
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25875G 5 g
D258725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25875G Supplier TCI America Supplier No. D25875G
Only null left

Chemical Identifiers

CAS 1455-77-2
Molecular Formula C2H5N5
Molecular Weight (g/mol) 99.097
MDL Number MFCD00005233
InChI Key PKWIYNIDEDLDCJ-UHFFFAOYSA-N
Synonym guanazole, 3,5-diamino-1,2,4-triazole, 4h-1,2,4-triazole-3,5-diamine, 3,5-diamino-1h-1,2,4-triazole, 3,5-diamino-s-triazole, s-triazole, 3,5-diamino, 1,2,4-triazolidine, 3,5-diimino, unii-i01twm5267, ccris 3745, 3-amino-1h-1,2,4-triazol-5-ylamine
PubChem CID 15078
ChEBI CHEBI:75425
IUPAC Name 1H-1,2,4-triazole-3,5-diamine
SMILES C1(=NC(=NN1)N)N

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 99.10
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Diamino-1,2,4-triazole
EINECSNumber (5)-3586
RTECSNumber XZ4535000
TSCA Yes