missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diamino-1,2,4-triazole, 98%

Catalog No. AAB2277514
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2277514 25 g
AAB2277506 5 g
AAB2277522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2277514 Supplier Thermo Scientific Chemicals Supplier No. B2277514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1455-77-2 | C2H5N5 | 99.097 g/mol

3,5-Diamino-1,2,4-triazole is used as an inhibitor of DNA synthesis. It also serves as an antitumor agents in the treatment of epigenetically-based diseases. It acts as a corrosion inhibitor for copper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Air sensitive. Incompatible with acids and oxidizing agents.

Chemical Identifiers

CAS 1455-77-2
Molecular Formula C2H5N5
Molecular Weight (g/mol) 99.097
MDL Number MFCD00005233
InChI Key PKWIYNIDEDLDCJ-UHFFFAOYSA-N
Synonym guanazole, 3,5-diamino-1,2,4-triazole, 4h-1,2,4-triazole-3,5-diamine, 3,5-diamino-1h-1,2,4-triazole, 3,5-diamino-s-triazole, s-triazole, 3,5-diamino, 1,2,4-triazolidine, 3,5-diimino, unii-i01twm5267, ccris 3745, 3-amino-1h-1,2,4-triazol-5-ylamine
PubChem CID 15078
ChEBI CHEBI:75425
IUPAC Name 1H-1,2,4-triazole-3,5-diamine
SMILES C1(=NC(=NN1)N)N

Specifications

Melting Point ∼205°C (decomposition)
Quantity 25 g
Beilstein 112467
Solubility Information Soluble in water.
Formula Weight 99.1
Percent Purity ≥98%
Chemical Name or Material 3,5-Diamino-1,2,4-triazole
EINECSNumber 215-937-2
RTECSNumber XZ4535000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only