missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di(trifluoromethyl)benzoic acid, Tech., Thermo Scientific™

Chemical Identifiers

CAS 725-89-3
Molecular Formula C9H4F6O2
Molecular Weight (g/mol) 258.12
MDL Number MFCD00000388
InChI Key HVFQJWGYVXKLTE-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzoic acid, 3,5-di trifluoromethyl benzoic acid, benzoic acid, 3,5-bis trifluoromethyl, bis 3,5-trifluoromethyl benzoic acid, mbt-boa, 3,5-bis-trifluoromethyl-benzoic acid, 3,5-bis-trifluoromethyl benzoic acid, 3.5-bis trifluoromethyl benzoic acid, pubchem2725
PubChem CID 12889
IUPAC Name 3,5-bis(trifluoromethyl)benzoic acid
SMILES OC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
DP01857DA
View Documents Promotion Details
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

725-89-3
258.12
HVFQJWGYVXKLTE-UHFFFAOYSA-N
12889
OC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
C9H4F6O2
MFCD00000388
3,5-bis trifluoromethyl benzoic acid, 3,5-di trifluoromethyl benzoic acid, benzoic acid, 3,5-bis trifluoromethyl, bis 3,5-trifluoromethyl benzoic acid, mbt-boa, 3,5-bis-trifluoromethyl-benzoic acid, 3,5-bis-trifluoromethyl benzoic acid, 3.5-bis trifluoromethyl benzoic acid, pubchem2725
3,5-bis(trifluoromethyl)benzoic acid

Specifications

725-89-3
100.0
Amber glass bottle
(CF3)2C6H3CO2H
3,5-bis trifluoromethyl benzoic acid, 3,5-di trifluoromethyl benzoic acid, benzoic acid, 3,5-bis trifluoromethyl, bis 3,5-trifluoromethyl benzoic acid, mbt-boa, 3,5-bis-trifluoromethyl-benzoic acid, 3,5-bis-trifluoromethyl benzoic acid, 3.5-bis trifluoromethyl benzoic acid, pubchem2725
OC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
258.12
258.12
Technical
80.0
97%
C9H4F6O2
MFCD00000388
HVFQJWGYVXKLTE-UHFFFAOYSA-N
3,5-bis(trifluoromethyl)benzoic acid
12889
Tech.
3, 5-Di(trifluoromethyl)benzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents