missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di(trifluoromethyl)benzoic acid, Tech., Thermo Scientific™

Catalog No. DP01857DA
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DP01857DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DP01857DA Supplier Thermo Scientific Chemicals Supplier No. DP01857DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 725-89-3
Molecular Formula C9H4F6O2
Molecular Weight (g/mol) 258.12
MDL Number MFCD00000388
InChI Key HVFQJWGYVXKLTE-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzoic acid, 3,5-di trifluoromethyl benzoic acid, benzoic acid, 3,5-bis trifluoromethyl, bis 3,5-trifluoromethyl benzoic acid, mbt-boa, 3,5-bis-trifluoromethyl-benzoic acid, 3,5-bis-trifluoromethyl benzoic acid, 3.5-bis trifluoromethyl benzoic acid, pubchem2725
PubChem CID 12889
IUPAC Name 3,5-bis(trifluoromethyl)benzoic acid
SMILES OC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

CAS Min % 80.0
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula (CF3)2C6H3CO2H
Quantity 1g
Formula Weight 258.12
Percent Purity Tech.
Grade Technical
Chemical Name or Material 3, 5-Di(trifluoromethyl)benzoic acid
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.