missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyltoluene 98.0+%, TCI America™

Catalog No. D34145G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34145G 5 g
D341425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34145G Supplier TCI America Supplier No. D34145G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15181-11-0
Molecular Formula C15H24
Molecular Weight (g/mol) 204.357
MDL Number MFCD00026300
InChI Key WIXDSJRJFDWTNY-UHFFFAOYSA-N
Synonym 3,5-di-tert-butyltoluene, 1,3-di-tert-butyl-5-methylbenzene, toluene, 3,5-di-tert-butyl, 3,5-bis tert-butyl toluene, benzene, 1,3-bis 1,1-dimethylethyl-5-methyl, 3,5-di-t-butyltoluene, 3,5-bis t-butyl-1-methylbenzene, 3,5-di-tert-butyl-1-methylbenzene, 1,3-bis tert-butyl-5-methylbenzene
PubChem CID 84819
IUPAC Name 1,3-ditert-butyl-5-methylbenzene
SMILES CC1=CC(=CC(=C1)C(C)(C)C)C(C)(C)C

Specifications

Color White-Yellow
Boiling Point 244°C
Quantity 5 g
Formula Weight 204.36
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3,5-Di-tert-butyltoluene
EINECSNumber (3)-3427
TSCA No