missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyltoluene, 98+%, Thermo Scientific Chemicals

$146.68 - $146.68

Chemical Identifiers

CAS 15181-11-0
Molecular Formula C15H24
Molecular Weight (g/mol) 204.357
MDL Number MFCD00026300
InChI Key WIXDSJRJFDWTNY-UHFFFAOYSA-N
Synonym 3,5-di-tert-butyltoluene, 1,3-di-tert-butyl-5-methylbenzene, toluene, 3,5-di-tert-butyl, 3,5-bis tert-butyl toluene, benzene, 1,3-bis 1,1-dimethylethyl-5-methyl, 3,5-di-t-butyltoluene, 3,5-bis t-butyl-1-methylbenzene, 3,5-di-tert-butyl-1-methylbenzene, 1,3-bis tert-butyl-5-methylbenzene
PubChem CID 84819
IUPAC Name 1,3-ditert-butyl-5-methylbenzene
SMILES CC1=CC(=CC(=C1)C(C)(C)C)C(C)(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1196114
View Documents
Thermo Scientific Chemicals
L1196114
25 g
Each for $146.68
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

15181-11-0
204.357
WIXDSJRJFDWTNY-UHFFFAOYSA-N
84819
CC1=CC(=CC(=C1)C(C)(C)C)C(C)(C)C
C15H24
MFCD00026300
3,5-di-tert-butyltoluene, 1,3-di-tert-butyl-5-methylbenzene, toluene, 3,5-di-tert-butyl, 3,5-bis tert-butyl toluene, benzene, 1,3-bis 1,1-dimethylethyl-5-methyl, 3,5-di-t-butyltoluene, 3,5-bis t-butyl-1-methylbenzene, 3,5-di-tert-butyl-1-methylbenzene, 1,3-bis tert-butyl-5-methylbenzene
1,3-ditert-butyl-5-methylbenzene

Specifications

15181-11-0
0.86
91°C (195°F)
1.49
25 g
3,5-di-tert-butyltoluene, 1,3-di-tert-butyl-5-methylbenzene, toluene, 3,5-di-tert-butyl, 3,5-bis tert-butyl toluene, benzene, 1,3-bis 1,1-dimethylethyl-5-methyl, 3,5-di-t-butyltoluene, 3,5-bis t-butyl-1-methylbenzene, 3,5-di-tert-butyl-1-methylbenzene, 1,3-bis tert-butyl-5-methylbenzene
CC1=CC(=CC(=C1)C(C)(C)C)C(C)(C)C
204.357
204.36
3,5-Di-tert-butyltoluene
31°C to 32°C
243°C to 244°C
C15H24
MFCD00026300
2043032
WIXDSJRJFDWTNY-UHFFFAOYSA-N
1,3-ditert-butyl-5-methylbenzene
84819
≥98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 239-230-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only