missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyltoluene, 98+%, Thermo Scientific Chemicals

Catalog No. AAL1196114
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1196114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1196114 Supplier Thermo Scientific Chemicals Supplier No. L1196114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 15181-11-0 | Molecular Formula: C15H24 | Molecular Weight: 204.357 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15181-11-0
Molecular Formula C15H24
Molecular Weight (g/mol) 204.357
MDL Number MFCD00026300
InChI Key WIXDSJRJFDWTNY-UHFFFAOYSA-N
Synonym 3,5-di-tert-butyltoluene, 1,3-di-tert-butyl-5-methylbenzene, toluene, 3,5-di-tert-butyl, 3,5-bis tert-butyl toluene, benzene, 1,3-bis 1,1-dimethylethyl-5-methyl, 3,5-di-t-butyltoluene, 3,5-bis t-butyl-1-methylbenzene, 3,5-di-tert-butyl-1-methylbenzene, 1,3-bis tert-butyl-5-methylbenzene
PubChem CID 84819
IUPAC Name 1,3-ditert-butyl-5-methylbenzene
SMILES CC1=CC(=CC(=C1)C(C)(C)C)C(C)(C)C

Specifications

Melting Point 31°C to 32°C
Density 0.86
Boiling Point 243°C to 244°C
Flash Point 91°C (195°F)
Refractive Index 1.49
Quantity 25 g
Beilstein 2043032
Formula Weight 204.36
Percent Purity ≥98%
Chemical Name or Material 3,5-Di-tert-butyltoluene
EINECSNumber 239-230-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only