missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butylsalicylaldehyde 98.0+%, TCI America™

Catalog No. p-7127423
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26465G 5 g
D264625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26465G Supplier TCI America Supplier No. D26465G
Only null left

Chemical Identifiers

CAS 37942-07-7
Molecular Formula C15H22O2
Molecular Weight (g/mol) 234.339
MDL Number MFCD00191998
InChI Key RRIQVLZDOZPJTH-UHFFFAOYSA-N
Synonym 3,5-di-tert-butyl-2-hydroxybenzaldehyde, 3,5-di-tert-butylsalicylaldehyde, 3,5-bis 1,1-dimethylethyl-2-hydroxy-benzaldehyde, benzaldehyde, 3,5-bis 1,1-dimethylethyl-2-hydroxy, 3,5-di-t-butylsalicylaldehyde, 3,5-di-tert-butyl-2-hydroxy-benzaldehyde, benzaldehyde, 2-hydroxy, 3,5-di t-butyl, 3,5-bis tert-butyl-2-hydroxybenzaldehyde, pubchem11247, acmc-1aete
PubChem CID 688023
IUPAC Name 3,5-ditert-butyl-2-hydroxybenzaldehyde
SMILES CC(C)(C)C1=CC(=C(C(=C1)C=O)O)C(C)(C)C

Specifications

Melting Point 63°C
Quantity 5 g
Formula Weight 234.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Di-tert-butylsalicylaldehyde