missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butylsalicaldehyde, 99%

Catalog No. AC458120050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458120050 5 g
AC458120250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC458120050 Supplier Thermo Scientific Chemicals Supplier No. 458120050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 37942-07-7 | C15H22O2 | 234.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 37942-07-7
Molecular Formula C15H22O2
Molecular Weight (g/mol) 234.34
InChI Key RRIQVLZDOZPJTH-UHFFFAOYSA-N
Synonym 3,5-di-tert-butyl-2-hydroxybenzaldehyde, 3,5-di-tert-butylsalicylaldehyde, 3,5-bis 1,1-dimethylethyl-2-hydroxy-benzaldehyde, benzaldehyde, 3,5-bis 1,1-dimethylethyl-2-hydroxy, 3,5-di-t-butylsalicylaldehyde, 3,5-di-tert-butyl-2-hydroxy-benzaldehyde, benzaldehyde, 2-hydroxy, 3,5-di t-butyl, 3,5-bis tert-butyl-2-hydroxybenzaldehyde, pubchem11247, acmc-1aete
PubChem CID 688023
IUPAC Name 3,5-ditert-butyl-2-hydroxybenzaldehyde
SMILES CC(C)(C)C1=CC(=C(C(=C1)C=O)O)C(C)(C)C

Specifications

Melting Point 61.0°C to 62.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Quantity 5 g
Formula Weight 234.34
Percent Purity 99%
Chemical Name or Material 3,5-Di-tert-butylsalicaldehyde
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only