missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butylbenzyl Alcohol 98.0+%, TCI America™

Catalog No. D26525G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26525G 5 g
D26521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26525G Supplier TCI America Supplier No. D26525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 77387-57-6
Molecular Formula C15H24O
Molecular Weight (g/mol) 220.36
MDL Number MFCD03844785
InChI Key VHYHRNYPVNFGNR-UHFFFAOYSA-N
PubChem CID 12660843
IUPAC Name (3,5-di-tert-butylphenyl)methanol
SMILES CC(C)(C)C1=CC(=CC(CO)=C1)C(C)(C)C

Specifications

Melting Point 61°C
Color White
Quantity 5 g
Formula Weight 220.36
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Di-tert-butylbenzyl Alcohol