missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butylbenzoic Acid 98.0+%, TCI America™

Catalog No. D26631G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26631G 1 g
D26635G 5 g
D266325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D26631G Supplier TCI America Supplier No. D26631G

Chemical Identifiers

CAS 16225-26-6
Molecular Formula C15H22O2
Molecular Weight (g/mol) 234.339
MDL Number MFCD00082727
InChI Key NCTSLPBQVXUAHR-UHFFFAOYSA-N
Synonym 3,5-di-tert-butylbenzoic acid, 3,5-di-t-butylbenzoic acid, 3,5-ditertbutylbenzoic acid, 3,5-di-tert-butylbenzoicacid, 3,5-bis tert-butyl benzoic acid, benzoic acid, 3,5-bis 1,1-dimethylethyl, pubchem4953, acmc-209do2, 3,5-di-tertbutylbenzoic acid, ksc494i2n
PubChem CID 85339
IUPAC Name 3,5-ditert-butylbenzoic acid
SMILES CC(C)(C)C1=CC(=CC(=C1)C(=O)O)C(C)(C)C

Specifications

Melting Point 173°C
Color White
Quantity 1 g
Formula Weight 234.34
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Di-tert-butylbenzoic Acid