missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butylbenzoic acid, 99%

Catalog No. AAL1238606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1238606 5 g
AAL1238603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1238606 Supplier Thermo Scientific Chemicals Supplier No. L1238606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16225-26-6 | C15H22O2 | 234.339 g/mol

3,5-Di-tert-butylbenzoic acid is used in the synthesis of 5,5-di-tert-butylanilinium sulfate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Di-tert-butylbenzoic acid is used in the synthesis of 5,5-di-tert-butylanilinium sulfate.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.

Chemical Identifiers

CAS 16225-26-6
Molecular Formula C15H22O2
Molecular Weight (g/mol) 234.339
MDL Number MFCD00082727
InChI Key NCTSLPBQVXUAHR-UHFFFAOYSA-N
Synonym 3,5-di-tert-butylbenzoic acid, 3,5-di-t-butylbenzoic acid, 3,5-ditertbutylbenzoic acid, 3,5-di-tert-butylbenzoicacid, 3,5-bis tert-butyl benzoic acid, benzoic acid, 3,5-bis 1,1-dimethylethyl, pubchem4953, acmc-209do2, 3,5-di-tertbutylbenzoic acid, ksc494i2n
PubChem CID 85339
IUPAC Name 3,5-ditert-butylbenzoic acid
SMILES CC(C)(C)C1=CC(=CC(=C1)C(=O)O)C(C)(C)C

Specifications

Melting Point 160°C to 162°C
Quantity 5 g
Beilstein 981248
Solubility Information Insoluble in water.
Formula Weight 234.34
Percent Purity 99%
Chemical Name or Material 3,5-Di-tert-butylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 240-350-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only