missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butylaniline 98.0+%, TCI America™

Catalog No. D25155G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25155G 5 g
D25151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25155G Supplier TCI America Supplier No. D25155G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2380-36-1
Molecular Formula C14H23N
Molecular Weight (g/mol) 205.345
MDL Number MFCD00134096
InChI Key MJKNHXCPGXUEDO-UHFFFAOYSA-N
Synonym 3,5-di-tert-butylaniline, 3,5-di-t-butylaniline, benzenamine, 3,5-bis 1,1-dimethylethyl, acmc-1cdyp, 3,5-ditert-butylphenylamine, 3,5-bis tert-butyl aniline, 3,5-di-tert-butylphenylamine, 3,5-bis tert-butyl phenylamine, mjknhxcpgxuedo-uhfffaoysa, 3,5-ditert-butyl-phenyl-amine
PubChem CID 75419
IUPAC Name 3,5-ditert-butylaniline
SMILES CC(C)(C)C1=CC(=CC(=C1)N)C(C)(C)C

Specifications

Melting Point 55°C
Quantity 5 g
Formula Weight 205.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Di-tert-butylaniline
EINECSNumber (3)-0129
TSCA No