missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyl-4-hydroxyphenylacetic acid, 98%

Catalog No. AAA1501506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1501506 5 g
AAA1501503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1501506 Supplier Thermo Scientific Chemicals Supplier No. A1501506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1611-03-6 | C16H24O3 | 264.365 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1611-03-6
Molecular Formula C16H24O3
Molecular Weight (g/mol) 264.365
MDL Number MFCD00156138
InChI Key QLMGIWHWWWXXME-UHFFFAOYSA-N
Synonym 3,5-di-tert-butyl-4-hydroxyphenylacetic acid, 3,5-di-tert-butyl-4-hydroxyphenyl acetic acid, 2-3,5-di tert-butyl-4-hydroxyphenyl acetic acid, 2-3,5-di-tert-butyl-4-hydroxyphenyl acetic acid, benzeneacetic acid,3,5-bis 1,1-dimethylethyl-4-hydroxy, acmc-1bvft, 4-hydroxy-3,5-di-t-butylphenylacetic acid, 3,5-di-t-butyl-4-hydroxyphenyl-acetic acid, 4-hydroxy-3,5-di-t-butyl-phenylacetic acid, 3,5-di-tert-butyl-4-hydroxybenzeneacetic acid
PubChem CID 2775110
IUPAC Name 2-(3,5-ditert-butyl-4-hydroxyphenyl)acetic acid
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)CC(=O)O

Specifications

Melting Point 154°C to 158°C
Quantity 5 g
Formula Weight 264.37
Percent Purity 98%
Chemical Name or Material 3,5-Di-tert-butyl-4-hydroxyphenylacetic acid
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only