missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyl-4-hydroxybenzyl alcohol, 97%

Catalog No. AAA1470906
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1470906 5 g
AAA1470914 25 g
AAA1470922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1470906 Supplier Thermo Scientific Chemicals Supplier No. A1470906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 88-26-6 | C15H24O2 | 236.355 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 88-26-6
Molecular Formula C15H24O2
Molecular Weight (g/mol) 236.355
MDL Number MFCD00017254
InChI Key HNURKXXMYARGAY-UHFFFAOYSA-N
Synonym 3,5-di-tert-butyl-4-hydroxybenzyl alcohol, 2,6-di-tert-butyl-4-hydroxymethyl phenol, 2,6-di-tert-butyl-4-hydroxymethylphenol, bht alcohol, antioxidant 754, ionox 100, ionox 100 antioxidant, 3,5-di-t-butyl-4-hydroxybenzyl alcohol, butylated hydroxymethylphenol, 2,6-di-t-butyl-4-hydroxymethylphenol
PubChem CID 6929
IUPAC Name 2,6-ditert-butyl-4-(hydroxymethyl)phenol
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)CO

Specifications

Melting Point 138°C to 141°C
Boiling Point 214°C (40 mmHg)
Quantity 5 g
Beilstein 2052291
Formula Weight 236.36
Percent Purity 97%
Chemical Name or Material 3,5-Di-tert-butyl-4-hydroxybenzyl alcohol
missing translation for 'healthHazard1' H315-H319-H335
missing translation for 'healthHazard2' GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-815-6
RTECSNumber DO0750000
TSCA Yes

RUO – Research Use Only