missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyl-4-hydroxybenzaldehyde, 98+%

CAS: 1620-98-0 | C15H22O2 | 234.339 g/mol

$61.26 - $533.12

Chemical Identifiers

CAS 1620-98-0
Molecular Formula C15H22O2
Molecular Weight (g/mol) 234.339
MDL Number MFCD00008826
InChI Key DOZRDZLFLOODMB-UHFFFAOYSA-N
Synonym 3,5-di-tert-butyl-4-hydroxybenzaldehyde, 3,5-di-t-butyl-4-hydroxybenzaldehyde, unii-95vti93vul, benzaldehyde, 3,5-bis 1,1-dimethylethyl-4-hydroxy, benzaldehyde, 3,5-di-tert-butyl-4-hydroxy, 95vti93vul, 3,5-ditert-butyl-4-hydroxy-benzaldehyde, 3,5-di tert-butyl-4-hydroxybenzaldehyde, 3,5-di-tert-butyl-4-hydroxy-benzaldehyde, 3,5-bis tert-butyl-4-hydroxybenzaldehyde
PubChem CID 73219
IUPAC Name 3,5-ditert-butyl-4-hydroxybenzaldehyde
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AAA1586406
View Documents
Thermo Scientific Chemicals
A1586406
5 g
Each for $61.26
Only null left
 
AAA1586414
View Documents
Thermo Scientific Chemicals
A1586414
25 g
Each for $192.42
Only null left
 
AAA1586422
View Documents
Thermo Scientific Chemicals
A1586422
100 g
Each for $533.12
Only null left
 
Description

Description

3,5-Di-tert-butyl-4-hydroxybenzaldehyde is employed as an OLED materials. It is also used as a pharmaceutical intermediate. The derivatives of this compound possess equipotent anti-inflammatory activities to indomethacin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Di-tert-butyl-4-hydroxybenzaldehyde is employed as an OLED materials. It is also used as a pharmaceutical intermediate. The derivatives of this compound possess equipotent anti-inflammatory activities to indomethacin.

Solubility
Soluble in hot methanol.

Notes
Air sensitive. Store at room temperature. Keep away from oxidizing agents.
Specifications

Chemical Identifiers

1620-98-0
234.339
DOZRDZLFLOODMB-UHFFFAOYSA-N
73219
CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C=O
C15H22O2
MFCD00008826
3,5-di-tert-butyl-4-hydroxybenzaldehyde, 3,5-di-t-butyl-4-hydroxybenzaldehyde, unii-95vti93vul, benzaldehyde, 3,5-bis 1,1-dimethylethyl-4-hydroxy, benzaldehyde, 3,5-di-tert-butyl-4-hydroxy, 95vti93vul, 3,5-ditert-butyl-4-hydroxy-benzaldehyde, 3,5-di tert-butyl-4-hydroxybenzaldehyde, 3,5-di-tert-butyl-4-hydroxy-benzaldehyde, 3,5-bis tert-butyl-4-hydroxybenzaldehyde
3,5-ditert-butyl-4-hydroxybenzaldehyde

Specifications

1620-98-0
C15H22O2
5 g
Air Sensitive
Soluble in hot methanol.
CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C=O
234.339
234.34
3,5-Di-tert-butyl-4-hydroxybenzaldehyde
187°C to 189°C
MFCD00008826
982526
3,5-di-tert-butyl-4-hydroxybenzaldehyde, 3,5-di-t-butyl-4-hydroxybenzaldehyde, unii-95vti93vul, benzaldehyde, 3,5-bis 1,1-dimethylethyl-4-hydroxy, benzaldehyde, 3,5-di-tert-butyl-4-hydroxy, 95vti93vul, 3,5-ditert-butyl-4-hydroxy-benzaldehyde, 3,5-di tert-butyl-4-hydroxybenzaldehyde, 3,5-di-tert-butyl-4-hydroxy-benzaldehyde, 3,5-bis tert-butyl-4-hydroxybenzaldehyde
DOZRDZLFLOODMB-UHFFFAOYSA-N
3,5-ditert-butyl-4-hydroxybenzaldehyde
73219
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 216-592-0

RTECSNumber : CU5610070

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only