Learn More
3,5-Di-tert-butyl-4-hydroxybenzaldehyde, 98+%
CAS: 1620-98-0 | C15H22O2 | 234.339 g/mol
Supplier: Thermo Scientific Chemicals A1586414
Description
3,5-Di-tert-butyl-4-hydroxybenzaldehyde is employed as an OLED materials. It is also used as a pharmaceutical intermediate. The derivatives of this compound possess equipotent anti-inflammatory activities to indomethacin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,5-Di-tert-butyl-4-hydroxybenzaldehyde is employed as an OLED materials. It is also used as a pharmaceutical intermediate. The derivatives of this compound possess equipotent anti-inflammatory activities to indomethacin.
Solubility
Soluble in hot methanol.
Notes
Air sensitive. Store at room temperature. Keep away from oxidizing agents.
Specifications
| 3,5-Di-tert-butyl-4-hydroxybenzaldehyde | |
| 187°C to 189°C | |
| MFCD00008826 | |
| 982526 | |
| 3,5-di-tert-butyl-4-hydroxybenzaldehyde, 3,5-di-t-butyl-4-hydroxybenzaldehyde, unii-95vti93vul, benzaldehyde, 3,5-bis 1,1-dimethylethyl-4-hydroxy, benzaldehyde, 3,5-di-tert-butyl-4-hydroxy, 95vti93vul, 3,5-ditert-butyl-4-hydroxy-benzaldehyde, 3,5-di tert-butyl-4-hydroxybenzaldehyde, 3,5-di-tert-butyl-4-hydroxy-benzaldehyde, 3,5-bis tert-butyl-4-hydroxybenzaldehyde | |
| DOZRDZLFLOODMB-UHFFFAOYSA-N | |
| 3,5-ditert-butyl-4-hydroxybenzaldehyde | |
| 73219 | |
| ≥98% |
| 1620-98-0 | |
| C15H22O2 | |
| 25 g | |
| Air Sensitive | |
| Soluble in hot methanol. | |
| CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C=O | |
| 234.339 | |
| 234.34 |
Chemical Identifiers
| 1620-98-0 | |
| 234.339 | |
| DOZRDZLFLOODMB-UHFFFAOYSA-N | |
| 73219 | |
| CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C=O |
| C15H22O2 | |
| MFCD00008826 | |
| 3,5-di-tert-butyl-4-hydroxybenzaldehyde, 3,5-di-t-butyl-4-hydroxybenzaldehyde, unii-95vti93vul, benzaldehyde, 3,5-bis 1,1-dimethylethyl-4-hydroxy, benzaldehyde, 3,5-di-tert-butyl-4-hydroxy, 95vti93vul, 3,5-ditert-butyl-4-hydroxy-benzaldehyde, 3,5-di tert-butyl-4-hydroxybenzaldehyde, 3,5-di-tert-butyl-4-hydroxy-benzaldehyde, 3,5-bis tert-butyl-4-hydroxybenzaldehyde | |
| 3,5-ditert-butyl-4-hydroxybenzaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 216-592-0
RTECSNumber : CU5610070
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only