missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)thiobenzamide, 97%

Catalog No. AAH2680903
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2680903 1 g
AAH26809MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2680903 Supplier Thermo Scientific Chemicals Supplier No. H2680903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 317319-15-6 | C9H5F6NS | 273.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 317319-15-6
Molecular Formula C9H5F6NS
Molecular Weight (g/mol) 273.20
MDL Number MFCD00278692
InChI Key OWYYZIVORPIBPZ-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl thiobenzamide, 3,5-di trifluoromethyl benzene-1-carbothioamide, 3,5-bis trifluoromethyl benzenecarbothioamide, 3,5-bis trifluoromethyl benzothioamide, benzenecarbothioamide,3,5-bis trifluoromethyl, acmc-20ands, maybridge1_003450, 3,5-bis-trifluoromethyl-thiobenzamide, 3,5-bis trifluoromethyl benzene-1-carbothioamide
PubChem CID 2806924
IUPAC Name 3,5-bis(trifluoromethyl)benzenecarbothioamide
SMILES NC(=S)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 112°C to 115°C
Quantity 1 g
Formula Weight 273.2
Percent Purity 97%
Chemical Name or Material 3,5-Bis(trifluoromethyl)thiobenzamide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only