missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)phenyldimethylchlorosilane, 95%

Catalog No. AAL1370903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1370903 1 g
AAL1370906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1370903 Supplier Thermo Scientific Chemicals Supplier No. L1370903

CAS: 732306-23-9 | C10H9ClF6Si | 306.707 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 732306-23-9
Molecular Formula C10H9ClF6Si
Molecular Weight (g/mol) 306.707
MDL Number MFCD01862186
InChI Key JZWDIKPZWMXPMG-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl phenyldimethylchlorosilane, 3,5-bis trifluoromethyl phenyl chloro dimethylsilane, 3,5-bis trifluoromethyl phenyl chloro dimethyl silane, 3,5-bis trifluoromethyl phenyl-chloro-dimethylsilane, 3,5-bis trifluoromethyl phenyl dimethylsilyl chloride, benzene,1-chlorodimethylsilyl-3,5-bis trifluoromethyl
PubChem CID 2783240
IUPAC Name [3,5-bis(trifluoromethyl)phenyl]-chloro-dimethylsilane
SMILES C[Si](C)(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)Cl

Specifications

Boiling Point 95°C to 96°C (30mmHg)
Quantity 1 g
UN Number UN3265
Formula Weight 306.71
Percent Purity 95%
Refractive Index 1.424
Sensitivity Moisture sensitive
Chemical Name or Material 3,5-Bis(trifluoromethyl)phenyldimethylchlorosilane
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only