CAS RN 732306-23-9
IUPAC Name:
[3,5-bis(trifluoromethyl)phenyl](chloro)dimethylsilane
Synonyms:
[3,5-bis(trifluoromethyl)phenyl](chloro)dimethylsilane
Molecular Weight (g/mol):
306.71
Molecular Formula:
C10H9ClF6Si
InChi Key:
JZWDIKPZWMXPMG-UHFFFAOYSA-N
SMILES:
C[Si](C)(Cl)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
3,5-Bis(trifluoromethyl)phenyldimethylchlorosilane, 95%
3,5-Bis(trifluoromethyl)phenyldimethylchlorosilane, 95%
CAS: 732306-23-9 Molecular Formula: C10H9ClF6Si Molecular Weight (g/mol): 306.707 MDL Number: MFCD01862186 InChI Key: JZWDIKPZWMXPMG-UHFFFAOYSA-N Synonym: 3,5-bis trifluoromethyl phenyldimethylchlorosilane,3,5-bis trifluoromethyl phenyl chloro dimethylsilane,3,5-bis trifluoromethyl phenyl chloro dimethyl silane,3,5-bis trifluoromethyl phenyl-chloro-dimethylsilane,3,5-bis trifluoromethyl phenyl dimethylsilyl chloride,benzene,1-chlorodimethylsilyl-3,5-bis trifluoromethyl PubChem CID: 2783240 IUPAC Name: [3,5-bis(trifluoromethyl)phenyl]-chloro-dimethylsilane SMILES: C[Si](C)(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)Cl