missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. B18861G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B18861G Supplier TCI America Supplier No. B18861G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 73852-19-4
Molecular Formula C8H5BF6O2
Molecular Weight (g/mol) 257.93
MDL Number MFCD00051850
InChI Key BPTABBGLHGBJQR-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl phenylboronic acid, 3,5-bis trifluoromethyl benzeneboronic acid, 3,5-bis trifluoromethyl phenyl boronic acid, 3,5-bis-trifluoromethylphenylboronic acid, 3,5-bis trifluoromethylphenyl boronic acid, 3,5-bis trifluoromethyl phenylboroic acid, btfpba, 3,5-bis trifluoromethyl phenyl boranediol
PubChem CID 156265
IUPAC Name [3,5-bis(trifluoromethyl)phenyl]boronic acid
SMILES OB(O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 220°C
Color White-Yellow
Quantity 1 g
Formula Weight 257.93
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Bis(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride)