missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)phenylacetic Acid 97.0+%, TCI America™

Catalog No. B19761G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19761G 1 g
B19765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19761G Supplier TCI America Supplier No. B19761G

Chemical Identifiers

CAS 85068-33-3
Molecular Formula C10H6F6O2
Molecular Weight (g/mol) 272.15
MDL Number MFCD00009908
InChI Key PAWSKKHEEYTXSA-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl phenylacetic acid, 2-3,5-bis trifluoromethyl phenyl acetic acid, 3,5-bis-trifluoromethylphenylacetic acid, mbt-paa, 2-3,5-di trifluoromethyl phenyl acetic acid, benzeneacetic acid, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl phenyl acetic acid, 3,5-di trifluoromethyl phenylacetic acid, 3,5-bis trifluoromethyl phenylaceticacid
PubChem CID 123587
IUPAC Name 2-[3,5-bis(trifluoromethyl)phenyl]acetic acid
SMILES OC(=O)CC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 125°C
Color White-Yellow
Quantity 1 g
Formula Weight 272.15
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Bis(trifluoromethyl)phenylacetic Acid