missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)phenyl Isothiocyanate 98.0+%, TCI America™

Catalog No. I056925G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I056925G 25 g
I05695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I056925G Supplier TCI America Supplier No. I056925G

Chemical Identifiers

CAS 23165-29-9
Molecular Formula C9H3F6NS
Molecular Weight (g/mol) 271.18
MDL Number MFCD00040838
InChI Key FXOSSGVJGGNASE-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-3,5-bis trifluoromethyl benzene, 3,5-bis trifluoromethyl phenylisothiocyanate, isothiocyanic acid 3,5-bis trifluoromethyl phenyl ester, 3,5-di trifluoromethyl phenyl isothiocyanate, 3,5-bis-trifluoromethyl phenyl isothiocyanate, 3,5-bis-trifluoromethylphenyl isothiocyanate, benzene, 1-isothiocyanato-3,5-bis trifluoromethyl, pubchem5019, acmc-209g1w
PubChem CID 2733395
IUPAC Name 1-isothiocyanato-3,5-bis(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC(=CC(=C1)N=C=S)C(F)(F)F

Specifications

Boiling Point 95°C
Quantity 25 g
UN Number 1760
Formula Weight 271.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Bis(trifluoromethyl)phenyl Isothiocyanate