missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)phenyl isothiocyanate, +99%, Thermo Scientific Chemicals

Catalog No. AC319520050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC319520050 5 g Glass Bottle
AC319520010 1 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC319520050 Supplier Thermo Scientific Chemicals Supplier No. 319520050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 23165-29-9 | C9H3F6NS | 271.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23165-29-9
Molecular Formula C9H3F6NS
Molecular Weight (g/mol) 271.18
MDL Number MFCD00040838
InChI Key FXOSSGVJGGNASE-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-3,5-bis trifluoromethyl benzene, 3,5-bis trifluoromethyl phenylisothiocyanate, isothiocyanic acid 3,5-bis trifluoromethyl phenyl ester, 3,5-di trifluoromethyl phenyl isothiocyanate, 3,5-bis-trifluoromethyl phenyl isothiocyanate, 3,5-bis-trifluoromethylphenyl isothiocyanate, benzene, 1-isothiocyanato-3,5-bis trifluoromethyl, pubchem5019, acmc-209g1w
PubChem CID 2733395
IUPAC Name 1-isothiocyanato-3,5-bis(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC(=CC(=C1)N=C=S)C(F)(F)F

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Yellow
Density 1.4850g/mL
Boiling Point 63.0°C (1.5 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Refractive Index 1.4989 to 1.5009
Linear Formula (CF3)2C6H3NCS
Quantity 5 g
Formula Weight 271.18
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material 3, 5-Bis(trifluoromethyl)phenyl isothiocyanate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face, hands and any exposed skin thoroughly after handling.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only